missing translation for 'onlineSavingsMsg'
Learn More

(3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one Hydrochloride, Mikromol™

Product Code. 18082110
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
18082110 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 18082110

Brand: Mikromol MM0252.100025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one Hydrochloride
CAS 119812-29-2
Analyte or Component Names (3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H20 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym Ondansetron Hydrochloride Dihydrate Imp. A (EP), Ondansetron Imp. A (EP), Ondansetron USP Related Compound A, Ondansetron USP RC A, (3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one Hydrochloride, Ondansetron Hydrochloride Dihydrate Imp. A (EP) as Hydrochloride, 3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one monohydrochloride, GR 37896A, BP Ondansetron Impurity A, Ph Eur Ondansetron Impurity A, USP Ondanestron Related Compound A, ondansetron mannich compound (GR 37896A), Ondansetron Related Compound A as Hydrochloride, Ondansetron Hydrochloride Dihydrate Impurity A as Hydrochloride, Ondansetron Impurity A as Hydrochloride
InChI Formula InChI=1S/C16H20N2O.ClH/c1-17(2)10-11-8-9-14-15(16(11)19)12-6-4-5-7-13(12)18(14)3;/h4-7,11H,8-10H2,1-3H3;1H
SMILES Cl.CN(C)CC1CCc2c(C1=O)c3ccccc3n2C
IUPAC Name 3-[(dimethylamino)methyl]-9-methyl-2,3-dihydro-1H-carbazol-4-one;hydrochloride
Molecular Weight (g/mol) 292.8
Quantity 25 mg
Formula Weight 292.1342 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.