missing translation for 'onlineSavingsMsg'
Learn More

(3RS)-3-[(1H-Imidazol-1-yl)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one (C-Demethylondansetron), Mikromol™

Product Code. 30049969
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30049969

Brand: Mikromol MM0252.070025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS)-3-[(1H-Imidazol-1-yl)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one (C-Demethylondansetron)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material C-Demethylondansetron
CAS 99614-03-6
Analyte or Component Names C-Demethylondansetron
Shipping Condition Room Temperature
Molecular Formula C17 H17 N3 O
Certifications/Compliance ISO 9001
Synonym Ondansetron Hydrochloride Dihydrate Imp. G (EP), Ondansetron Imp. G (EP), (3RS)-3-[(1H-Imidazol-1-yl)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, C-Demethylondansetron, Ondansetron Hydrochloride Dihydrate Impurity G, Ondansetron Impurity G
InChI Formula InChI=1S/C17H17N3O/c1-19-14-5-3-2-4-13(14)16-15(19)7-6-12(17(16)21)10-20-9-8-18-11-20/h2-5,8-9,11-12H,6-7,10H2,1H3
SMILES Cn1c2CCC(Cn3ccnc3)C(=O)c2c4ccccc14
IUPAC Name 3-(imidazol-1-ylmethyl)-9-methyl-2,3-dihydro-1H-carbazol-4-one
Molecular Weight (g/mol) 279.34
Quantity 25 mg
Formula Weight 279.1372 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.