missing translation for 'onlineSavingsMsg'
Learn More

(3R,S)-Oxidosqualene (>90%), TRC

Product Code. 30416043
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416043 2.5 mg
30415203 1 mg
30413494 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416043 Supplier Toronto Research Chemicals Supplier No. TRCO8468152.5MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7200-26-2
Molecular Formula C30H50O
Molecular Weight (g/mol) 426.72
Synonym 2,2-Dimethyl-3-[(3E,7E,11E,15E)-3,7,12,16,20-pentamethyl-3,7,11,15,19-heneicosapentaen-1-yl]oxirane, (all-E)-22,23-Epoxy-2,6,10,15,19,23-hexamethyl-2,6,10,14,18-tetracosapentaene, 2,2-Dimethyl-3-[(3E,7E,11E,15E)-3,7,12,16,20-pentamethyl-3,7,11,15,19-heneicosapentaenyl]oxirane, (R,S)-Squalene 2,3-epoxide, (RS)-2,3-Epoxy-2,3-dihydrosqualene, (±)-2,3-Epoxysqualene, (±)-Squalene oxide, 2,3-Epoxysqualene, 2,3-Oxidosqualene, Squalene 2,3-epoxide, Squalene 2,3-oxide, Squalene epoxide, Squalene oxide
IUPAC Name 2,2-dimethyl-3-[(3E,7E,11E,15E)-3,7,12,16,20-pentamethylhenicosa-3,7,11,15,19-pentaenyl]oxirane
SMILES CC(=CCC\C(=C\CC\C(=C\CC\C=C(/C)\CC\C=C(/C)\CCC1OC1(C)C)\C)\C)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C30H50O/c1-24(2)14-11-17-27(5)20-12-18-25(3)15-9-10-16-26(4)19-13-21-28(6)22-23-29-30(7,8)31-29/h14-16,20-21,29H,9-13,17-19,22-23H2,1-8H3/b25-15+,26-16+,27-20+,28-21+
Formula Weight 426.3862
Chemical Name or Material (3R,S)-Oxidosqualene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.