missing translation for 'onlineSavingsMsg'
Learn More

(3R,5R,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid Calcium Salt, TRC

Product Code. 30362153
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362153 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362153 Supplier Toronto Research Chemicals Supplier No. TRCC988855100MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,5R,6E)-Pitavastatin Calcium
CAS 254452-96-5
Molecular Formula 2 C25 H23 F N O4 . Ca
Quantity 100 mg
Synonym 6-Heptenoic acid, 7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-, calcium salt (2:1), (3R,5R,6E)-, 5-epi-NK-104, (3R,5R,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic acid hemicalcium salt, (3R,5R,6E)-Pit
InChI Formula InChI=1S/2C25H24FNO4.Ca/c2*26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31;/h2*1-4,7-12,16,18-19,28-29H,5-6,13-14H2,(H,30,31);/q;;+2/p-2/b2*12-11+;/t2*18-,19+;/m00./s1
SMILES [Ca+2].O[C@H](C[C@@H](O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4)CC(=O)[O-].O[C@H](C[C@@H](O)\C=C\c5c(nc6ccccc6c5c7ccc(F)cc7)C8CC8)CC(=O)[O-]
IUPAC Name calcium;(E,3R,5R)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol) 880.984
Formula Weight 880.285
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.