missing translation for 'onlineSavingsMsg'
Learn More

(3R,4S,5S)-4-Hydroxy-3-(pentan-3-yloxy)-5-(tosyloxy)cyclohex-1-enecarboxylic Acid, TRC

Product Code. 30407881
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.50mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407881 5 mg 5mg
30404549 25 mg 25mg
30392288 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407881 Supplier Toronto Research Chemicals Supplier No. TRCH9399905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,4S,5S)-4-Hydroxy-3-(pentan-3-yloxy)-5-(tosyloxy)cyclohex-1-enecarboxylic Acid
Analyte or Component Names (3R,4S,5S)-4-Hydroxy-3-(pentan-3-yloxy)-5-(tosyloxy)cyclohex-1-enecarboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C21 H30 O7 S
InChI Formula InChI=1S/C21H30O7S/c1-5-16(6-2)27-18-12-15(21(23)26-7-3)13-19(20(18)22)28-29(24,25)17-10-8-14(4)9-11-17/h8-12,16,18-20,22H,5-7,13H2,1-4H3/t18-,19+,20+/m1/s1
Solution Type Neat
SMILES CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@H](O)[C@H](C1)OS(=O)(=O)c2ccc(C)cc2
IUPAC Name ethyl (3R,4S,5S)-4-hydroxy-5-(4-methylphenyl)sulfonyloxy-3-pentan-3-yloxycyclohexene-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 426.524
Quantity 5 mg
Formula Weight 426.171
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.