missing translation for 'onlineSavingsMsg'
Learn More

(3R,4S,5R)-4-Hydroxy-5-[(1S)-1-hydroxy-2-methylpropyl]-3-methyl-2-pyrrolidinone-5-carboxylic Acid, TRC

Product Code. 30402473
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402473 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402473 Supplier Toronto Research Chemicals Supplier No. TRCH9431205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,4S,5R)-4-Hydroxy-5-[(1S)-1-hydroxy-2-methylpropyl]-3-met
CAS 145451-97-4
Molecular Formula C10 H17 N O5
Quantity 5 mg
InChI Formula InChI=1S/C10H17NO5/c1-4(2)6(12)10(9(15)16)7(13)5(3)8(14)11-10/h4-7,12-13H,1-3H3,(H,11,14)(H,15,16)/t5-,6+,7+,10-/m1/s1
SMILES CC(C)[C@H](O)[C@]1(NC(=O)[C@H](C)[C@@H]1O)C(=O)O
IUPAC Name (2R,3S,4R)-3-hydroxy-2-[(1S)-1-hydroxy-2-methylpropyl]-4-methyl-5-oxopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 231.25
Formula Weight 231.1107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.