missing translation for 'onlineSavingsMsg'
Learn More

(3R,4R,4aS,7R,8S,8aR)-Hexahydro-3-hydroxy-4,7-dimethyl-8-(3-oxobutyl)-1,2-benzodioxin-8a(3H)-carboxaldehyde, TRC

Product Code. 30397215
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397215 500 mg
30394893 10 mg
30400301 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397215 Supplier Toronto Research Chemicals Supplier No. TRCH294045500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149588-86-3
Molecular Formula C15H24O5
Molecular Weight (g/mol) 284.35
Synonym [3R-(3α,4β,4aα,7α,8β,8aβ)]-Hexahydro-3-hydroxy-4,7-dimethyl-8-(3-oxobutyl)-1,2-benzodioxin-8a(3H)-carboxaldehyde
IUPAC Name (3R,4R,4aS,7R,8S,8aR)-3-hydroxy-4,7-dimethyl-8-(3-oxobutyl)-4,4a,5,6,7,8-hexahydro-3H-benzo[c][1,2]dioxine-8a-carbaldehyde
SMILES C[C@@H]1CC[C@H]2[C@H]([C@@H](OO[C@@]2([C@H]1CCC(=O)C)C=O)O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H24O5/c1-9-4-6-13-11(3)14(18)19-20-15(13,8-16)12(9)7-5-10(2)17/h8-9,11-14,18H,4-7H2,1-3H3/t9-,11-,12+,13+,14-,15-/m1/s1
Formula Weight 284.1624
Chemical Name or Material (3R,4R,4aS,7R,8S,8aR)-Hexahydro-3-hydroxy-4,7-dimethyl-8-(3-oxobutyl)-1,2-benzodioxin-8a(3H)-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.