missing translation for 'onlineSavingsMsg'
Learn More

(3R,4R,2'S)-3-Hexyl-4-(2'-benzyloxypentadec-1'-yl) Oxetan-2-one, TRC

Product Code. 30390750
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390750 5 mg 5mg
30402394 10 mg 10mg
30389938 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390750 Supplier Toronto Research Chemicals Supplier No. TRCH2956285MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,4R,2'S)-3-Hexyl-4-(2'-benzyloxypentadec-1'-yl) Oxetan-2-one
Analyte or Component Names (3R,4R,2'S)-3-Hexyl-4-(2'-benzyloxypentadec-1'-yl) Oxetan-2-one
Shipping Condition Room Temperature
Molecular Formula C31 H52 O3
Synonym (3R,4R)-4-((S)-2-(Benzyloxy)pentadecyl)-3-hexyloxetan-2-one
InChI Formula InChI=1S/C31H52O3/c1-3-5-7-9-10-11-12-13-14-15-19-23-28(33-26-27-21-17-16-18-22-27)25-30-29(31(32)34-30)24-20-8-6-4-2/h16-18,21-22,28-30H,3-15,19-20,23-26H2,1-2H3/t28-,29+,30+/m0/s1
Solution Type Neat
SMILES CCCCCCCCCCCCC[C@@H](C[C@H]1OC(=O)[C@@H]1CCCCCC)OCc2ccccc2
IUPAC Name (3R,4R)-3-hexyl-4-[(2S)-2-phenylmethoxypentadecyl]oxetan-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 472.743
Quantity 5 mg
Formula Weight 472.392
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.