missing translation for 'onlineSavingsMsg'
Learn More

(3R)-3-Hydroxybutyric Acid-1-13C, TRC

Product Code. 30393717
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393717 5 mg
30397176 100 mg
30398576 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393717 Supplier Toronto Research Chemicals Supplier No. TRCH8330285MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C313CH8O3
Molecular Weight (g/mol) 105.1
Synonym (3R)-3-Hydroxybutanoic Acid-1-13C, (R)-3-Hydroxybutanoic Acid-1-13C, D-(-)-3-Hydroxybutyric Acid-1-13C, (R)-(-)-3-Hydroxybutyrate-1-13C, KTX 0101-1-13C, (R)-3-Hydroxybutyrate-1-13C, D-β-Hydroxybutyrate-1-13C, R-3-Hydroxybutyrate-1-13C
SMILES O[C@H](C)C[13C](O)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C4H8O3/c1-3(5)2-4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m1/s1/i4+1
Chemical Name or Material (3R)-3-Hydroxybutyric Acid-1-13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.