missing translation for 'onlineSavingsMsg'
Learn More

(3R)-3,6-Dihydro-2H-1,4-thiazine-3,5-dicarboxylic Acid 3-Ethyl Ester, TRC

Product Code. 30386766
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386766 50 mg -
30369997 500 mg 500mg
30374993 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386766 Supplier Toronto Research Chemicals Supplier No. TRCD45066050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944115-20-2
Molecular Formula C8H11NO4S
Molecular Weight (g/mol) 217.24
Synonym (5R)-5-(Ethoxycarbonyl)-5,6-dihydro-2H-1,4-thiazine-3-carboxylic Acid, Lanthionine Ketimine-R-5-ethyl Ester
SMILES OC(C1=CSC[C@H](C(OCC)=O)N1)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H11NO4S/c1-2-13-8(12)6-4-14-3-5(9-6)7(10)11/h6H,2-4H2,1H3,(H,10,11)/t6-/m0/s1
Formula Weight 217.0409
Chemical Name or Material (3R)-3,6-Dihydro-2H-1,4-thiazine-3,5-dicarboxylic Acid 3-Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.