missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 3PO

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
3PO is an originally reported to inhibit PFKFB3 (IC50 = 25 μM). More recent reports failed to demonstrate activity in a PFKFB3 kinase assay. Reduces glycolytic flux and suppresses glucose uptake. Inhibits endothelial cell proliferation and causes G2/M cell cycle arrest in vitro. Attenuates vessel sprouting and tumor growth in vivo. Amplifies the antiangiogenic effect of VEGFR blockade.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | 3PO |
| Purity | >99% |
| Molecular Weight (g/mol) | 210.23 |
| Inhibitors | 3PO |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction