Learn More
3M™ Spectacle Kits for 7000 Series Full Facepiece Respirators
Use with 3M™ Full Facepieces 7000 Series.
Brand: 3M™ 7000088361
Code : NEW
Additional Details : Weight : 0.06650kg
Description
- Permits the wearer to use corrective lenses and to make position adjustment that is most comfortable.
- 3M Catalog# 7925/7925-AM
Specifications
Spectacle Kit | |
Demolition, facility maintenance, grinding, sanding, welding | |
Full facepiece 7000 series | |
7925/7925-AM |