missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scotch-Brite™ 3M Scotch double sided tape, 19mm x 33m
Description
Double sided transparent tape for long terms use. Ideal for craft projects. Permanent, durable and moisture resistant. No backing strip.
Size: 19mm x 33m
Size: 19mm x 33m
Specifications
Specifications
| Length (Metric) | 33 m |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?