missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ S-Series Hood with Faceseal, Integrated Head Suspension

Versaflo™ S-Series Hood with integrated suspension provides head, face, neck, and shoulder coverage as well as providing eye and face protection from liquid splash and low energy flying particles and dust.

Brand:  3M™ 7100011287

82.50 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.



Code : S8

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11771153

  • £138.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

  • Integrated head suspension provides head, face, neck and shoulder coverage.
  • Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F).
  • Suitable for applications which require frequent replacement of the entire headtop assembly or those where there is less soiling of the outer fabric.
  • No assembly required
  • Ready to use straight out of the box.
Specifications

Specifications

1000
White
Hood with Faceseal
Assembly, facility maintenance, machine operation, painting, sanding
Coated Polycarbonate
1 Ea.
NIOSH
Polyurethane-coated Knitted Polyamide
Small/Medium
GVP, Breathe-Easy™, V Series, T-300, T-600, T-800
S-533 S
10 x 9.63 x 8.19 in. (25.4 x 24.46 x 20.8 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers