Learn More
3M™ Protective Coverall 4570
3M™ Protective Coverall 4570 helps protect against a range of chemical jets and sprays. This chemical protective suit has been certified to Type 3/4/5/6 levels of protection.
Brand:  3M™ 7000089688_OLD
Product Code. 15749847
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
3M™ Protective Coverall 4570 offers chemical hold out and mechanical strength for workers in environments with chemical exposure. The suit helps protect against hazardous dusts (Type 5), light liquid splashes (Type 6), low pressure liquid sprays (Type 4) and high pressure liquid jets (Type 3). It is also certified to offer protection against radioactive particles (EN 1073-2) and (EN 14126). This suit achieves the highest class performance on all tests in EN 14126 and passes ASTM F1670 and F1671 for helping to protect against biological contaminants.
  • A Versatile Protective Suit for many Applications
  • Protective coverall helps protect against a range of chemical jets and sprays
  • Heavy-duty fabric with a soft anti-static treated inner layer offers protection and convenience
  • Stitched seams are sealed with a 5-layer chemically protective tape
  • Two storm flaps create a double seal for extra protection
  • Large thumb loops and zipper pulls offer convenience while wearing gloves
  • Knit polyester cuffs for user comfort
  • Anti-static, low-linting, microporous
  • Elastic around the head for secure fit
  • Elastic around the face to ensure secure fit
  • Elastic at the calf to provide closure around the top of the footwear for protection against contaminants
  • Elastic around the ankle for protection against contaminants and enhanced fit
  • Universal size with ties to provide a good fit for a wide range of users
  • Lightweight and comfortable to wear
  • 3M Catalog# 4570-2XL, 4570-3XL, 4570-4XL, 4570-L, 4570-M, 4570-XL