missing translation for 'onlineSavingsMsg'
Learn More

3M™ TR-300 and TR-600 PAPR Battery Packs

Product Code. 11703906
Change view
Click to view available options
For Use With (Equipment):
Versaflo TR-300 PAPR Series
Versaflo TR-600 PAPR Series
Battery Life:
4 to 6 hr.
8 to 12 hr.
8 to 18 hr.
Color:
Black
Blue, Gray
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Battery Life Color unitSize
11703906 Versaflo TR-300 PAPR Series 8 to 12 hr. Black Each
11793896 Versaflo TR-300 PAPR Series 4 to 6 hr. Black Each
15549305 Versaflo TR-600 PAPR Series - Blue, Gray Each
15589255 Versaflo TR-600 PAPR Series 8 to 18 hr. Blue, Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11703906 Supplier 3M™ Supplier No. 7100005643

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Choose from a selection of lithium ion rechargeable batteries for the TR-300 or TR-600 PAPR systems.

  • Rechargeable battery packs
  • Lithium Ion
  • Long run time for filters and cartridges

Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product

Compliance

NIOSH approved.

TRUSTED_SUSTAINABILITY

Specifications

Battery Life 8 to 12 hr.
Battery Type Lithium-ion
Color Black
Product Type Battery Pack
For Use With (Application) Supplying power for PAPR systems
For Use With (Equipment) Versaflo TR-300 PAPR Series
Model TR-332
Recharge Time <3.5 hr.
Weight (English) 2.4 lb.
Weight (Metric) 1.077 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.