missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Overboot Cover 441

Product Code. 15779037
Change view
Click to view available options
:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15779037 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779037 Supplier 3M™ Supplier No. 7000089719

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The 3M™ Disposable boot covers are designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.

The 3M™ Overboot Covers 440 are a reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Offered as an accessory, the overboots are made from a microporous laminate material with an anti-static treatment.
  • Covers are designed with ties and elastic at the calf to help ensure a good fit
  • Distinctive unisex design
  • Integrated sideshields with adjustable lens angle and temple lengths
  • Vertically vented browgard minimizes fogging
TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Material Microporous Fabric
Coating Microporous Film Laminate
Color White
Disposable Single-use
Closure Type Elastic Top
For Use With (Application) Assembly, Cleaning, Machine Operations
Resistance Dry Particulates, Fluid
Product Type Boot Covers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.