Learn More
3M™ Disposable Protective Lab Coat, Series 4440
Offers a barrier against non-hazardous particles and liquid splashes. The 3M Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.
Brand: 3M™ 7000089696
This item is not returnable.
View return policy
Description
- Breathable, lightweight material
- Designed for light duty applications
- Offers a barrier against non-hazardous particles and liquid splashes
- Zip fastener
- Anti-static treated
- Knitted cuffs
- 3M Catalog# 4440-2XL, 4440-4XL, 4440-M, 4440-XL