missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3M™ Comply™ Indicator Tape

Product Code. 11741605
Click to view available options
Dimensions (L x W):
55 m x 18 mm
55 m x 24 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11741605

Brand: Thermo Scientific™ 1322V24

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.

As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.
  • Lead-free steam indicator tape is designed for use on all wraps
  • Reliable achesive adheres to packaging material
  • Sustainable, solvent-free manufacturing proces
  • Does not require hazardous waste disposal
  • Redesigned packaging uses up to 30% less plastic
  • Convenient, individually wrapped, easy-to-open rolls
  • Lead free and no added latex
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets NF EN 867-1 Class A, ISO 11140-1 Class 1
Color Beige
Dimensions (L x W) 55 m x 24 mm
For Use With (Application) Sterilization verification
For Use With (Equipment) Autoclave
Length (Metric) 55 m
Product Type Indicator Tape
Features Lead-free, aggressive, pressure sensitive adhesive
Quantity 20 Pk.
Sterilization Indicator Yes
Sterilization Method Steam
Width (Metric) Exterior 24 mm
Show More Show Less
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.