missing translation for 'onlineSavingsMsg'
Learn More

3H,5H-Octamethyltetrasiloxane, 96%

Product Code. 11343856
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343856 25 g 25g
11333856 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343856 Supplier Thermo Scientific Alfa Aesar Supplier No. L16956.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16066-09-4 | C8H24O3Si4 | 280.617 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16066-09-4
Molecular Formula C8H24O3Si4
Molecular Weight (g/mol) 280.617
MDL Number MFCD00053664
InChI Key OHSYWAVRSCQMHG-UHFFFAOYSA-N
Synonym 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc
PubChem CID 6327318
IUPAC Name methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon
SMILES C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 169°C
Quantity 25 g
Flash Point 78°C (172°F)
Formula Weight 282.64
Percent Purity 96%
Refractive Index 1.386
Sensitivity Moisture sensitive
Density 0.863
Chemical Name or Material 3H,5H-Octamethyltetrasiloxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.