missing translation for 'onlineSavingsMsg'
Learn More

3H-1,2-Dithiole-3-thione, TRC

Product Code. 30385018
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385018 100 mg
30375920 10 mg
30371641 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385018 Supplier Toronto Research Chemicals Supplier No. TRCD494365100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-25-8
Molecular Formula C3 H2 S3
Molecular Weight (g/mol) 134.24
IUPAC Name dithiole-3-thione
SMILES S=C1SSC=C1

Specifikationer

Quantity 100 mg
InChI Formula InChI=1S/C3H2S3/c4-3-1-2-5-6-3/h1-2H
Formula Weight 133.9319
Chemical Name or Material 3H-1,2-Dithiole-3-thione
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.