missing translation for 'onlineSavingsMsg'
Learn More

3H-1,2-Benzodithiol-one 1,1-dioxide, 98%, Thermo Scientific Chemicals

Product Code. 11364576
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66304-01-6
Molecular Formula C7H4O3S2
Molecular Weight (g/mol) 200.226
MDL Number MFCD00132960
InChI Key JUDOLRSMWHVKGX-UHFFFAOYSA-N
PubChem CID 3009847
IUPAC Name 1,1-dioxo-1$l^{6},2-benzodithiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)SS2(=O)=O

Specifications

Melting Point 102°C to 107°C
Quantity 250 mg
UN Number UN3335
Beilstein 4180332
Synonym 3h-1,2-benzodithiol-3-one 1,1-dioxide, beaucage reagent, 3h-1,2-benzodithiol-3-one-1,1-dioxide, 3h-benzo c 1,2 dithiol-3-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one, 1,1-dioxide, 1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one, 3h-1,2-benzodithiol-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one1,1-dioxide, 1??,2-benzodithiole-1,1,3-trione, acmc-1b2sk
Formula Weight 200.24
Percent Purity 99%
Chemical Name or Material 3H-1,2-Benzodithiol-one 1,1-dioxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'