missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film

Single-use tank liners reduce cleaning steps common to re-usable tanks.

Brand:  Thermo Scientific™ B100363-I

Additional Details : Weight : 7.00000kg

Product Code. 15949786

  • POA / 20 units
Estimated Shipment: 13-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications

Specifications

200 L
Store at room temperature.
Individually packed in a header bag
Irradiated
HyPerforma™
ASI™ 26
Polyethylene
55.9 cm x 91.4 cm (22 in. x 36 in.)
20 per case
Room Temperature
0 ports
No Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers