missing translation for 'onlineSavingsMsg'
Learn More

3Beta-Hydroxychol-5-enoic Acid Methyl Ester, TRC

Product Code. 30402007
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402007 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402007 Supplier Toronto Research Chemicals Supplier No. TRCH91780010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3Beta-Hydroxychol-5-enoic Acid Methyl Ester
CAS 20231-57-6
Molecular Formula C25H40O3
Quantity 10 mg
Synonym L 601920-0, Methyl 3β-hydroxychol-5-en-24-oate, Methyl 3β-hydroxychol-5-enoate, Methyl Δ5-3β-Hydroxycholanoate, Methyl-3β-hydroxycholenate, NSC 84140
InChI Formula InChI=1S/C25H40O3/c1-16(5-10-23(27)28-4)20-8-9-21-19-7-6-17-15-18(26)11-13-24(17,2)22(19)12-14-25(20,21)3/h6,16,18-22,26H,5,7-15H2,1-4H3/t16-,18+,19+,20-,21+,22+,24+,25-/m1/s1
SMILES CO[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCC(=O)OC
IUPAC Name methyl (4R)-4-[(3S,8S,9S,10R,13R,14S,17R)-3-methoxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 388.58
Formula Weight 402.3134
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.