missing translation for 'onlineSavingsMsg'
Learn More

3Beta-Hydroxy Progesterone, TRC

Product Code. 30389977
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389977 500 mg -
30391302 1 g -
30396618 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389977 Supplier Toronto Research Chemicals Supplier No. TRCH939040500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 566-66-5
Molecular Formula C21H32O2
Molecular Weight (g/mol) 316.48
Synonym Pregn-4-en-20-one, 3β-hydroxy- (8CI), (3β)-3-Hydroxypregn-4-en-20-one, 3-Hydroxy-4-pregnen-20-one, 3β-Hydroxypregn-4-en-20-one, 4-Pregnen-3β-ol-20-one, δ4-Pregnene-3β-ol-20-one
IUPAC Name 1-[(3S,8S,9S,10R,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,15-19,23H,4-11H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1
Formula Weight 316.2402
Chemical Name or Material 3Beta-Hydroxy Progesterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.