missing translation for 'onlineSavingsMsg'
Learn More

3Beta-Hydroxy Pravastatin Lactone (>90%), TRC

Product Code. 30400784
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400784 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400784 Supplier Toronto Research Chemicals Supplier No. TRCH9523051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3beta-Hydroxy Pravastatin Lactone
CAS 87984-67-6
Molecular Formula C23 H34 O6
Quantity 1 mg
Synonym Butanoic acid, 2-methyl-, (1S,6R,7R,8S,8aR)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)-, Butanoic acid, 2-methyl-, 1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1α(R*),6β,7β,8β(2S*,4S*),8aβ]]- (9CI), (2S)-2-Methylbutanoic acid (1S,6R,7R,8S,8aR)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, 3β-Hydroxy pravastatin lactone, 3β-Hydroxypravastatin lactone, 6β-Hydroxy-iso-compactin
InChI Formula InChI=1S/C23H34O6/c1-4-13(2)23(27)29-20-7-5-6-15-10-19(25)14(3)18(22(15)20)9-8-17-11-16(24)12-21(26)28-17/h5-6,10,13-14,16-20,22,24-25H,4,7-9,11-12H2,1-3H3/t13-,14+,16+,17+,18-,19-,20-,22-/m0/s1
SMILES CC[C@H](C)C(=O)O[C@H]1CC=CC2=C[C@H](O)[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
IUPAC Name [(1S,6R,7R,8S,8aR)-6-hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-7-methyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
Molecular Weight (g/mol) 406.51
Formula Weight 406.2355
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.