missing translation for 'onlineSavingsMsg'
Learn More

3Beta-Glycocholic Acid, TRC

Product Code. 30395112
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395112 10 mg
30393334 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395112 Supplier Toronto Research Chemicals Supplier No. TRCG64135510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107589-98-0
Molecular Formula C26H43NO6
Molecular Weight (g/mol) 465.62
Synonym N-[(3β,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]glycine, 3β-N-Choloylglycine, 3β-Glycoreductodehydrocholic Acid, 3β,7α,12α-Trihydroxy-5β-cholanic αcid-24-glycine, 3β-Cholylglycine, 3β-Glycine Cholate
IUPAC Name 2-[[(4R)-4-[(3S,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
SMILES O[C@@H]1C[C@]2([H])C[C@@H](O)CC[C@]2(C)[C@]3([H])[C@]1([H])[C@@]4([H])[C@]([C@@H]([C@H](C)CCC(NCC(O)=O)=O)CC4)(C)[C@@H](O)C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H43NO6/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33)/t14-,15+,16+,17-,18+,19+,20-,21+,24+,25+,26-/m1/s1
Formula Weight 465.309
Chemical Name or Material 3beta-Glycocholic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.