missing translation for 'onlineSavingsMsg'
Learn More

3Beta,5Beta-Dihydroxy Drospirenone-d4 Ring-opened Alcohol Impurity, TRC

Product Code. 30384937
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384937 5 mg 5mg
30378329 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30384937 Supplier Toronto Research Chemicals Supplier No. TRCD4527925MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3beta,5beta-Dihydroxy Drospirenone-d4 Ring-opened Alcohol Impurity
Molecular Formula C24H34D4O4
Quantity 5 mg
Synonym (3S,5R,6R,7R,8R,9S,10R,13S,14S,15S,16S,17S)-Octadecahydro-17-(3-hydroxypropyl)-10,13-dimethyl-5H-dicyclopropa[6,7:15,16]cyclopenta[a]phenanthrene-3,5,17-triol-d4, [3S-(3α,5α,6β,7β,8α,9β,10α,13α,14β,15β,16β,17α)]-Octadecahydro-17-(3-hydroxypropyl)-10,13-dimethyl-5H-dicyclopropa[6,7:15,16]cyclopenta[a]phenanthrene-3,5,17-triol-d4, 17-(3-Hydroxypropyl)-6β,7β:15β,16β-dimethyleneandrostane-3β,5β,17β-triol-d4
InChI Formula InChI=1S/C24H38O4/c1-21-7-4-13(26)12-24(21,28)17-10-14(17)19-16(21)5-8-22(2)20(19)15-11-18(15)23(22,27)6-3-9-25/h13-20,25-28H,3-12H2,1-2H3/t13-,14-,15+,16-,17+,18-,19+,20-,21?,22?,23-,24+/m0/s1/i3D2,6D2
SMILES O[C@]1(C([2H])([2H])C([2H])([2H])CO)[C@@]2(C)CC[C@@]3([H])[C@@]([C@]2([H])[C@H]4[C@@H]1C4)([H])[C@@H]5[C@@H](C5)[C@]6(O)C[C@@H](O)CC[C@@]63C
IUPAC Name (2S,4aR,4bS,6aS,7S,7aS,8aS,8bS,8cR,8dR,9aR,9bR)-7-(3-hydroxypropyl-1,1,2,2-d4)-4a,6a-dimethyloctadecahydro-9bH-cyclopropa[4,5]cyclopenta[1,2-a]cyclopropa[l]phenanthrene-2,7,9b-triol
Molecular Weight (g/mol) 394.58
Formula Weight 394.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.