missing translation for 'onlineSavingsMsg'
Learn More

(3beta,5alpha)-4,4-Dimethylcholesta-8,14,24-trien-3-ol(Contain up to 15% ∆25 isomer), TRC

Product Code. 30384550
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384550 5 mg 5mg
30382415 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384550 Supplier Toronto Research Chemicals Supplier No. TRCD4717255MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3beta,5alpha)-4,4-Dimethylcholesta-8,14,24-trien-3-ol
CAS 64284-64-6
Molecular Formula C29H46O
Quantity 5 mg
Synonym 4,4-Dimethyl-5α-cholesta-8,14,24-trien-3β-ol, FF-MAS, Follicular Fluid Meiosis-activating Sterol
InChI Formula InChI=1S/C29H46O/c1-19(2)9-8-10-20(3)22-12-13-23-21-11-14-25-27(4,5)26(30)16-18-29(25,7)24(21)15-17-28(22,23)6/h9,13,20,22,25-26,30H,8,10-12,14-18H2,1-7H3/t20-,22-,25+,26+,28-,29-/m1/s1
SMILES C[C@H](CCC=C(C)C)[C@H]1CC=C2[C@@]1(CCC3=C2CC[C@@H]4[C@@]3(CC[C@@H](C4(C)C)O)C)C
IUPAC Name (3S,5R,10S,13R,17R)-4,4,10,13-tetramethyl-17-[(2R)-6-methylhept-5-en-2-yl]-1,2,3,5,6,7,11,12,16,17-decahydrocyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 410.67
Formula Weight 410.3549
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.