missing translation for 'onlineSavingsMsg'
Learn More

(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Acetate (Mutilin 14-Acetate), Mikromol™

Product Code. 30052055
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052055 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052055 Supplier Mikromol Supplier No. MM0394.13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Acetate (Mutilin 14-Acetate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Acetate (Mutilin 14-Acetate)
CAS 31771-36-5
Analyte or Component Names (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Acetate (Mutilin 14-Acetate)
Shipping Condition Room Temperature
Molecular Formula C22 H34 O4
Certifications/Compliance ISO 9001
Synonym (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Acetate, Tiamulin Hydrogen Fumarate Imp. J (EP), Mutilin 14-Acetate, Tiamulin Imp. J (EP), Tiamulin Hydrogen Fumarate Impurity J, Tiamulin Impurity J
InChI Formula InChI=1S/C22H34O4/c1-7-20(5)12-17(26-15(4)23)21(6)13(2)8-10-22(14(3)19(20)25)11-9-16(24)18(21)22/h7,13-14,17-19,25H,1,8-12H2,2-6H3/t13-,14+,17-,18+,19+,20-,21+,22+/m1/s1
SMILES C[C@@H]1CC[C@@]23CCC(=O)[C@H]2[C@]1(C)[C@@H](C[C@@](C)(C=C)[C@@H](O)[C@@H]3C)OC(=O)C
IUPAC Name (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Acetate
Molecular Weight (g/mol) 362.5
Quantity 50 mg
Formula Weight 362.2457 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.