missing translation for 'onlineSavingsMsg'
Learn More

[3aS-(3aAlpha,4Beta,6aAlpha)]-Hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-Thieno[3,4-d]imidazole-4-pentanoic Acid Phenylmethyl Ester, TRC

Product Code. 30401428
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401428 25 mg 25mg
30388818 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401428 Supplier Toronto Research Chemicals Supplier No. TRCH29433025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [3aS-(3aalpha,4beta,6aalpha)]-Hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-Thieno[3,4-d]imidazole-4-pentanoic Acid Phenylmethyl Ester
CAS 179532-60-6
Molecular Formula C31H34N2O3S
Quantity 25 mg
Synonym (3aS,4S,6aR)-Hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-thieno[3,4-d]imidazole-4-pentanoic Acid Phenylmethyl Ester
InChI Formula InChI=1S/C31H34N2O3S/c34-29(36-22-26-16-8-3-9-17-26)19-11-10-18-28-30-27(23-37-28)32(20-24-12-4-1-5-13-24)31(35)33(30)21-25-14-6-2-7-15-25/h1-9,12-17,27-28,30H,10-11,18-23H2/t27-,28-,30-/m0/s1
SMILES O=C(CCCC[C@@H]1SC[C@H]2[C@@H]1N(Cc3ccccc3)C(=O)N2Cc4ccccc4)OCc5ccccc5
IUPAC Name benzyl 5-[(3aS,4S,6aR)-1,3-dibenzyl-2-oxo-3a,4,6,6a-tetrahydrothieno[3,4-d]imidazol-4-yl]pentanoate
Molecular Weight (g/mol) 514.68
Formula Weight 514.229
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.