missing translation for 'onlineSavingsMsg'
Learn More

(3aR,4R,6S,6aS)-4-[[(1,1-Dimethylethoxy)carbonyl]amino]-3-(1-ethylpropyl)-3a,5,6,6a-tetrahydro-4H-cyclopent[d]isoxazole-6-carboxylic Acid Methyl Ester, TRC

Product Code. 30381527
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381527 1 g
30381990 5 g
30377107 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381527 Supplier Toronto Research Chemicals Supplier No. TRCD4728901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 229613-93-8
Molecular Formula C18H30N2O5
Molecular Weight (g/mol) 354.44
Synonym Peramivir Impurity 1
SMILES CCC(CC)C1=NO[C@@H]2[C@H](C[C@@H](NC(=O)OC(C)(C)C)[C@H]12)C(=O)OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H30N2O5/c1-7-10(8-2)14-13-12(19-17(22)24-18(3,4)5)9-11(16(21)23-6)15(13)25-20-14/h10-13,15H,7-9H2,1-6H3,(H,19,22)/t11-,12+,13+,15+/m0/s1
Formula Weight 354.2155
Chemical Name or Material (3aR,4R,6S,6aS)-4-(((1,1-Dimethylethoxy)carbonyl)amino)-3-(1-ethylpropyl)-3a,5,6,6a-tetrahydro-4H-cyclopent(d)isoxazole-6-carboxylic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.