missing translation for 'onlineSavingsMsg'
Learn More

(3aR,4R,5R,6aS)-Hexahydro-2-oxo-4-[(1E)-3-oxo-1-octen-1-yl]-2H-cyclopenta[b]furan-5-yl Ester [1,1’-Biphenyl]-4-carboxylic Acid, TRC

Product Code. 30395559
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395559 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395559 Supplier Toronto Research Chemicals Supplier No. TRCH29437510MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material (3aR,4R,5R,6aS)-Hexahydro-2-oxo-4-[(1E)-3-oxo-1-octen-1-yl]-2H-cyclopenta[b]furan-5-yl Ester [1,1’-Biphenyl]-4-carboxylic Acid
CAS 31753-00-1
Molecular Formula C28H30O5
Quantity 10 mg
Synonym 2H-Cyclopenta[b]furan, [1,1'-biphenyl]-4-carboxylic Acid Deriv, [3aα,4α(E),5β,6aα]-(-)-[1,1'-Biphenyl]-4-carboxylic Acid, Hexahydro-2-oxo-4-(3-oxo-1-octenyl)-2H-cyclopenta[b]furan-5-yl Ester
InChI Formula InChI=1S/C28H30O5/c1-2-3-5-10-22(29)15-16-23-24-17-27(30)32-26(24)18-25(23)33-28(31)21-13-11-20(12-14-21)19-8-6-4-7-9-19/h4,6-9,11-16,23-26H,2-3,5,10,17-18H2,1H3/b16-15+/t23-,24-,25-,26+/m1/s1
SMILES CCCCCC(/C=C/[C@H]1[C@@H](C[C@@H]2OC(C[C@H]12)=O)OC(C3=CC=C(C=C3)C4=CC=CC=C4)=O)=O
IUPAC Name (3aR,4R,5R,6aS)-2-oxo-4-((E)-3-oxooct-1-en-1-yl)hexahydro-2H-cyclopenta[b]furan-5-yl [1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol) 446.53
Formula Weight 446.21
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.