missing translation for 'onlineSavingsMsg'
Learn More

(3aR,4R,5R,6aS)-4-[3-(Ethyleneketal)decanyl]hexahydro-5-hydroxy-2H-cyclopenta[b]furan-2-one-d15, TRC

Product Code. 30382921
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382921 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382921 Supplier Toronto Research Chemicals Supplier No. TRCE91778710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3aR,4R,5R,6aS)-4-[3-(Ethyleneketal)decanyl]hexahydro-5-hydroxy-2H-cyclopenta[b]furan-2-one-d15
CAS 1217783-38-4
Molecular Formula C19H17D15O5
Quantity 10 mg
Synonym [3aR-(3aalpha,4alpha,5beta,6aalpha)]-4-[2-(2-Heptyl-1,3-dioxolan-2-yl)ethyl]hexahydro-5-hydroxy-2H-cyclopenta[b]furan-2-one-d15
InChI Formula InChI=1S/C19H32O5/c1-2-3-4-5-6-8-19(22-10-11-23-19)9-7-14-15-12-18(21)24-17(15)13-16(14)20/h14-17,20H,2-13H2,1H3/t14-,15-,16-,17+/m1/s1/i1D3,2D2,3D2,4D2,5D2,6D2,8D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1(CC[C@H]2[C@H](O)C[C@@H]3OC(=O)C[C@H]23)OCCO1
IUPAC Name (3aR,4R,5R,6aS)-5-hydroxy-4-[2-[2-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecadeuterioheptyl)-1,3-dioxolan-2-yl]ethyl]-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one
Molecular Weight (g/mol) 355.55
Formula Weight 355.3191
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.