missing translation for 'onlineSavingsMsg'
Learn More

3alpha-Hydroxy-4beta-(piperidin-1-yl)-5alpha-androstan-17-one, Mikromol™

Product Code. 30049288
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30049288 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30049288

Brand: Mikromol MM1620.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3alpha-Hydroxy-4beta-(piperidin-1-yl)-5alpha-androstan-17-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3alpha-Hydroxy-4beta-(piperidin-1-yl)-5alpha-androstan-17-one
CAS 1142951-28-7
Analyte or Component Names 3alpha-Hydroxy-4beta-(piperidin-1-yl)-5alpha-androstan-17-one
Shipping Condition Room Temperature
Molecular Formula C24 H39 N O2
Certifications/Compliance ISO 17025
Synonym (3α,4β,5α)-3-Hydroxy-4-(1-piperidinyl)androstan-17-one
InChI Formula InChI=1S/C24H39NO2/c1-23-13-11-20(26)22(25-14-4-3-5-15-25)19(23)7-6-16-17-8-9-21(27)24(17,2)12-10-18(16)23/h16-20,22,26H,3-15H2,1-2H3/t16-,17-,18-,19-,20+,22+,23+,24-/m0/s1
SMILES C[C@]12CC[C@@H](O)[C@@H]([C@@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC4=O)N5CCCCC5
IUPAC Name (3R,4R,5R,8R,9S,10R,13S,14S)-3-hydroxy-10,13-dimethyl-4-piperidin-1-yl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 373.57
Quantity 25 mg
Formula Weight 373.2981 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.