missing translation for 'onlineSavingsMsg'
Learn More

3alpha,7alpha,12alpha-Trihydroxy-5beta-cholan-24-oic Acid (Cholic Acid), Mikromol™

Product Code. 17455469
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17455469 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17455469 Supplier Mikromol Supplier No. MM0835.020250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3alpha,7alpha,12alpha-Trihydroxy-5beta-cholan-24-oic Acid (Cholic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cholic Acid
CAS 81-25-4
Analyte or Component Names Cholic Acid
Shipping Condition Room Temperature
Molecular Formula C24 H40 O5
Certifications/Compliance ISO 17034
Synonym Cholic Acid, (3α,5β,7α,12α)-3,7,12-Trihydroxycholan-24-oic acid, Cholic acid, 17β-[1-Methyl-3-carboxypropyl]etiocholane-3α,7α,12α-triol, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanoic acid, 3α,7α,12α-Trihydroxy-β-cholanic acid, 3α,7α,12α-Trihydroxycholanic acid, 5β-Cholanic acid-3α,7α,12α-triol, 5β-Cholic acid, Cholalic acid, Cholalin, Colalin, E 1000, NSC-6135, Chenodeoxycholic Acid Impurity B, Ursodeoxycholic Acid Impurity B
InChI Formula InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 408.57
Quantity 250 mg
Formula Weight 408.2876 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.