missing translation for 'onlineSavingsMsg'
Learn More

(3Alpha,5Beta,7Alpha)-7-Hydroxy-3-(sulfooxy)-cholan-24-oic Acid Disodium Salt, TRC

Product Code. 30391909
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391909 10 mg 10mg
30395444 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391909 Supplier Toronto Research Chemicals Supplier No. TRCH95402510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60237-34-5
Molecular Formula C24H38Na2O7S
Molecular Weight (g/mol) 516.6
Synonym Cholan-24-oic acid, 7-hydroxy-3-(sulfooxy)-, disodium salt, (3α,5β,7α)- (9CI), (3α,5β,7α)-7-Hydroxy-3-(sulfooxy)-cholan-24-oic acid disodium salt, Disodium 3-sulfochenodeoxycholate, Chenodeoxycholic acid 3-sulfate disodium salt, Chenodeoxycholic acid 3α-sulfate disodium salt, Chenodeoxycholic acid 3-O-sulfate disodium salt
IUPAC Name disodium;(4R)-4-[(3R,5R,7R,8R,9S,10S,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-sulfonatooxy-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [H][C@@]12C[C@H](OS(=O)(O[Na])=O)CC[C@]1(C)[C@]3([H])[C@@]([C@@](CC[C@]4([H])[C@@H](CCC(O[Na])=O)C)([H])[C@]4(C)CC3)([H])[C@H](O)C2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H40O7S.2Na/c1-14(4-7-21(26)27)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(31-32(28,29)30)12-15(23)13-20(22)25;;/h14-20,22,25H,4-13H2,1-3H3,(H,26,27)(H,28,29,30);;/q;2*+1/p-2/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-;;/m1../s1
Formula Weight 516.2134
Percent Purity >95%
Chemical Name or Material (3alpha,5beta,7alpha)-7-Hydroxy-3-(sulfooxy)-cholan-24-oic Acid Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.