missing translation for 'onlineSavingsMsg'
Learn More

3Alpha,12Alpha-Dihydroxy-5Beta-chol-9(11)-enic Acid, TRC

Product Code. 30386705
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386705 10 mg
30371071 25 mg
30376927 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386705 Supplier Toronto Research Chemicals Supplier No. TRCD45280810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24637-46-5
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym Chol-9(11)-en-24-oic acid, 3,12-dihydroxy-, (3α,5β,12α)- (9CI), 5β-Chol-9(11)-enic acid, 3α,12α-dihydroxy- (6CI), 3α,12α-Dihydroxy-5β-chol-9(11)-enic acid
IUPAC Name (4R)-4-[(3R,5R,8S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)C3=C[C@H](O)[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H38O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h13-19,21,25-26H,4-12H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,21+,23+,24-/m1/s1
Formula Weight 390.277
Percent Purity >95%
Chemical Name or Material 3alpha,12alpha-Dihydroxy-5beta-chol-9(11)-enic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.