missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3a)-Allopregnanolone, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Allopregnanolone |
| CAS | 516-54-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21 H34 O2 |
| Quantity | 10 mg |
| Synonym | (+)-3alpha,-Hydroxy-5alpha,-pregnan-20-one, (3alpha,,5alpha,)-3-Hydroxypregnan-20-one, 3alpha,,5alpha,-THP, 3alpha,,5alpha,-Tetrahydroprogesterone, 3alpha,-Hydroxy-5alpha,-dihydroprogesterone, 3alpha,-Hydroxy-5alpha,-pregnan-20-one, 3alpha,-Hydroxy-5alpha,-pregnane-20-one, 5alpha,-Pregnan-3alpha,-ol-20-one, 5alpha,-Pregnane-3alpha,-ol-20-one, Allopregnan-3alpha,-ol-20-one, Allopregnanolone, Allotetrahydroprogesterone |
| InChI Formula | InChI=1S/C20H32O2/c1-19-9-7-15(22)11-13(19)3-5-16-17-6-4-14(12-21)20(17,2)10-8-18(16)19/h12-18,22H,3-11H2,1-2H3/t13-,14+,15+,16-,17-,18-,19-,20+/m0/s1 |
| SMILES | C[C@@]12[C@@]3([C@]([C@]4([C@](C)(CC3)[C@@H](C(C)=O)CC4)[H])(CC[C@]1(C[C@H](O)CC2)[H])[H])[H] |
| IUPAC Name | 1-[(3R,5S,8R,9S,10S,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone |
| Molecular Weight (g/mol) | 318.49 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction