missing translation for 'onlineSavingsMsg'
Learn More

(3a,5b,7a)-3,7-Dihydroxy-12-oxocholan-24-oic Acid Methyl Ester, TRC

Product Code. 30377943
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377943 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377943 Supplier Toronto Research Chemicals Supplier No. TRCD453895100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3alpha,5beta,7alpha)-3,7-Dihydroxy-12-oxocholan-24-oic Acid Methyl Ester
CAS 10538-64-4
Molecular Formula C25H40O5
Quantity 100 mg
Synonym 3alpha,7alpha-Dihydroxy-12-oxo-5beta-cholan-24-oic Acid Methyl Ester, 3alpha,7alpha-Dihydroxy-12-oxo-5beta-cholanic Acid Methyl Ester, 12-Ketochenodeoxycholic Acid Methyl Ester, 12-Monoketocholic Acid Methyl Ester, 3alpha,7alpha-Dihydroxy-12-oxo-5beta-cholanic Acid Methyl Ester, Methyl 3alpha,7alpha-Dihydroxy-12-oxo-5beta-cholan-24-oate
InChI Formula InChI=1S/C25H40O5/c1-14(5-8-22(29)30-4)17-6-7-18-23-19(13-21(28)25(17,18)3)24(2)10-9-16(26)11-15(24)12-20(23)27/h14-20,23,26-27H,5-13H2,1-4H3/t14-,15+,16-,17-,18+,19+,20-,23+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC(=O)[C@]12C
Molecular Weight (g/mol) 420.58
Formula Weight 420.2876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.