Learn More
Greiner Bio-One 384-well Small Volume™ HiBase Polystyrene Microplates
Product Code. p-7075409
Click to view available options
Color:
Black
Sterility:
Sterile
No. per Case:
40
Unit Size:
Pack of 40
Product Code. 13017993
Brand: Greiner Bio-One 784077
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features round wells with a conical geometry. Greiner Bio-One 384-Well Small Volume™ HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Working volume: 4μL - 25μL
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Sterility Sterile
No. per Case 40
No. per Pack 10