missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 384-well Polystyrene Flat Bottom Microplates

Product Code. 10073301
Click to view available options
Binding Property:
High Binding
No Binding
Lid:
With Lid
Without Lid
No. per Pack:
1
8
10
Unit Size:
Pack of 32
Pack of 40
This item is not returnable. View return policy

Product Code. 10073301

Brand: Greiner Bio-One 781186

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Free of detectable DNase, RNase, human DNA. Greiner Bio-One 384 Well Polystyrene Flat Bottom Microplates are compatible with standard equipment and automated systems.

  • Manufactured under DIN ISO 9001 guidelines
  • Non-Pyrogenic
  • Number of wells are quadrupled
  • Well-to-well spacing is 4.5mm
  • Compatible with standard equipment and automated systems
  • Rounded square well profile

Service Plan

No

TRUSTED_SUSTAINABILITY

Specifications

Binding Property No Binding
Format Standard
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 32
No. per Pack 8
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.