missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 384-Well Polypropylene Plates
Feature higher well density for economical use of expensive reagents
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 7.06000kg
Description
- Ideal for storage of compounds and nucleic acid libraries
- Chemical-resistant
- Withstand temperatures from -80° to +80°C (-112° to +176°F)
- Conform to ANSI™-standard footprint for optimal use in automated handling and liquid-dispensing systems
Specifications
ANSI | |
16 x 24 | |
Polypropylene | |
Non-sterile |
Standard | |
Red | |
384 | |
For storage of compounds and nucleic acid libraries |