Learn More
Greiner Bio-One 384-well Polypropylene Microplates
Product Code. 10407102
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 100
Product Code. Color unitSize
10407102 Natural Pack of 100
1 options
Product Code. 10407102 Supplier Greiner Bio-One Supplier No. 781280
Only null left
Add to basket
Add to basket
Manufactured with polypropylene for optimum chemical resistance and high temperature tolerance. Greiner Bio-One™ 384 Well Polypropylene Microplates provide four times the number of wells in a 96 well standard microplate while retaining external dimensions compatible with standard equipment and automated systems.
  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Well-to-well spacing: 4.5mm
  • Low biomolecular binding characteristics
  • Resistant to many standard laboratory chemicals, such as DMSO 
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN ISO 9001
Certified DNase/RNase and human DNA free
Format Standard
Array 16 x 24
Packaging Inner packs of 10
Color Natural
Product Type Genomic
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 10 to 130 μL
Sterility Non-sterile
Total Volume (Metric) Well 138 μL
Well Shape Conical Bottom (V-well)
Quantity 100/Cs.