missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WebSeal 384-Well Non-Coated Plastic Microplates, U-Shaped Well

For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 384 well microplates.

Brand:  Thermo Scientific™ 60180-P106

Code : NEW

Additional Details : Weight : 0.95000kg

 View more versions of this product

Product Code. 15102709

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 384-Well Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:

  • Pharmaceutical and industrial QA/QC
  • High-throughput screening (HTS)
  • Sample collection and storage
  • Combinatorial chemistry
  • Liquid phase assays

Chemically and Thermally Resistant Polypropylene

  • Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
  • Will tolerate temperatures from −80° C to 121°C
  • GC-tested to ensure low extractables

Broad Instrument Compatibility

  • SBS and ANSI standard footprint design

Well Formats to Suit Every Application

  • Shallow well and deep well formats
  • U- and V- bottom wells ensure optimal sample recovery
Specifications

Specifications

Standard
Microplate
384
Non-sterile
58 μL
25 Microplates
16 x 24
Polypropylene
2 to 35 μL
Non-coated
Round Bottom (U-well)
14.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.