Learn More
BRAND™ 384-Well PCR Plates
Made with polypropylene material, these plates are resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems.
Brand: BRAND™ 781347
Code : 03
Additional Details : Weight : 0.95000kg
Description
- Non-sterile, 0.3mL BRAND™ 384-well deep well plate has a square bottom and is manufactured from high-quality virgin polypropylene
- Excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, sample storage, and more
- Plate is resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems
- Plate withstands temperatures as low as -121ºC, making it an excellent choice for cold-room applications and cryo-storage
- PP plate is autoclavable at 250ºF (121ºC) for 20 minutes
- Wells are labeled in a standard alpha-numeric pattern to simplify sample identification, and have notched corners to facilitate orientation
Specifications
Standard | |
Autoclavable | |
Full | |
384 | |
50/Pk. |
16 x 24 | |
Full skirt | |
Polypropylene | |
qPCR |