missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 384-Well Microtiter Microplates

Product Code. 10160485
Change view
Click to view available options
Color:
Clear
Sterility:
Non-sterile
Lid:
Without Lid
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Lid unitSize
10160485 Clear Non-sterile Without Lid Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10160485 Supplier Thermo Scientific™ Supplier No. 95040000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Excellent for a number of applications.

These microplates share the same footprint as 96-well versions.

  • Square wells
  • Design allows for miniaturization of assays and other applications
  • Use for high-volume DNA library manipulation and solid-phase binding

Use these microplates for the following application:

  • Clear plates—colorimetric applications
  • White plates—luminometric/fluorometric applications
  • Black plates—fluorometric applications

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Packaging 50 Cases
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
For Use With (Application) For high throughput screening, PCR, mutagenicity/mutational spectra assays
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.