missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Low Flange Polystyrene Microplates

For use with fluorescent assays with detection microplate reader. Corning™ 384-Well Low Flange Polystyrene Microplates have low background fluorescence, minimal light scatter and reduced crosstalk.

Brand:  Corning™ 3769

Additional Details : Weight : 21.90000kg

Product Code. 15772907

  • £3178.12 / Pack of 100
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

Specifications

Specifications

Standard
0.06 cm2
White, Clear Bottom
110.5 μL
Non-sterile
Flat Bottom (F-well)
20/Pk.
11.6 mm
With Lid
16 x 24
Polystyrene
384
Non-binding
100/Cs.
127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Top/bottom: 3.63/2.82 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers