missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Bar-Coded Microplate
Shop All Corning Products

Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.06 cm2 |
| Array | 16 x 24 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 25 to 80 μL |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?