missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porvair Sciences™ 384-Well High Binding Polystyrene Microplates
Description
- Square wells maximise liquid volume per well (120μL/well)
- White plate prevents cross talk in light emitting assays
- Black plate gives low autofluorescence background
- Tissue culture treated plates for anchorage dependent cells
- Total well volume 120μL
- Dimensions [w x d x h] 3.75mm x 3.75mm x 11.4mm (well), 85.5mm x 127.5mm x 14.8mm (plate)
- 24 x 16 well array
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Material | Polystyrene |
| No. of Wells | 384 |
| Total Volume (Metric) Well | 120 μL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?